Thermo Scientific Chemicals

Benzethonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 105380025
2.5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Benzethonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: 105380025
2.5 kg, Plastic drum, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 105380025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-54-0
IUPAC Name
benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
Molecular Formula
C27H42ClNO2
InChI Key
UREZNYTWGJKWBI-UHFFFAOYSA-M
SMILES
[Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
Appearance (Form)
Powder or crystalline powder
Loss on drying
=<5 % (105°C, 4 h)
Melting point
158°C to 163°C
Appearance (Color)
White to almost white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text