Thermo Scientific Chemicals

alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals

Catalog number: 105530250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals

Catalog number: 105530250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 105530250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
441-38-3
IUPAC Name
(2E)-2-(N-hydroxyimino)-1,2-diphenylethan-1-ol
Molecular Formula
C14H13NO2
InChI Key
WAKHLWOJMHVUJC-FYWRMAATNA-N
SMILES
O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Melting point
150°C to 155°C
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text