alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals
alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 105535000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS441-38-3
IUPAC Name(2E)-2-(N-hydroxyimino)-1,2-diphenylethan-1-ol
Molecular FormulaC14H13NO2
InChI KeyWAKHLWOJMHVUJC-FYWRMAATNA-N
SMILESO\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point150°C to 155°C
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only