Benzophenone, 99%, pure
Benzophenone, 99%, pure
Benzophenone, 99%, pure
Benzophenone, 99%, pure
Thermo Scientific Chemicals

Benzophenone, 99%, pure

CAS: 119-61-9 | C13H10O | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
1 kg
Packaging:
View all
Plastic bottle
Catalog number 105565000
Price (USD)
41.65
Special offer
Online exclusive
49.30
Save 7.65 (16%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
49.30
Save 7.65 (16%)
Each
Add to cart
Chemical Identifiers
CAS119-61-9
IUPAC Namediphenylmethanone
Molecular FormulaC13H10O
InChI KeyRWCCWEUUXYIKHB-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
GC>=98.5 %
Appearance (Color)White to off-white
Melting point47°C to 49°C
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473822Certificate of AnalysisJun 25, 2025105560010, 105565000, 105560050
A0473820Certificate of AnalysisJun 25, 2025105560010, 105565000, 105560050
A0473821Certificate of AnalysisJun 25, 2025105560010, 105565000, 105560050
A0472111Certificate of AnalysisMay 17, 2025105560010, 105565000, 105560050
A0466051Certificate of AnalysisSep 10, 2024105560010, 105565000, 105560050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection