Thermo Scientific Chemicals

2,1,3-Benzothiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: 105660250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,1,3-Benzothiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: 105660250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 105660250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
273-13-2
IUPAC Name
2,1,3-benzothiadiazole
Molecular Formula
C6H4N2S
InChI Key
PDQRQJVPEFGVRK-UHFFFAOYSA-N
SMILES
S1N=C2C=CC=CC2=N1
Appearance (Color)
Colorless to white to light yellow
Appearance (Form)
Low melting solid
Infrared spectrum
Conforms
Melting point
42°C to 47°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text