Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

DMEM, high glucose, GlutaMAX™ Supplement

Catalog number: 10566032
Have Questions?
Gibco™

DMEM, high glucose, GlutaMAX™ Supplement

Catalog number: 10566032
Have Questions?
Catalog Number
10566032
Unit Size
5 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
105660325 L
-
10566016500 mL
-
1056602410 x 500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This DMEM is modified as follows:
WithWithout
• High Glucose• Sodium Pyruvate
• GlutaMAX™ 
• Phenol Red 


The complete formulation is available.

Using DMEM
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Specifications

Cell Line
HeLa, 293, Cos-7, and PC-12
Cell Type
Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Classification
Animal Origin-free
Concentration
1 X
Form
Liquid
Product Type
DMEM (Dulbecco's Modified Eagle Medium)
Serum Level
Standard Serum Supplementation
Shelf Life
12 Months From Date of Manufacture
Sterility
Sterile-filtered
With Additives
High Glucose, GlutaMAX, Phenol Red
Without Additives
No HEPES, No Sodium Pyruvate
Manufacturing Quality
cGMP-compliant under the ISO 13485 standard
Product Line
Gibco™, GlutaMAX™, SynerGy™
Quantity
5 L
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text