Benzoxazole, 97+%, Thermo Scientific Chemicals
Benzoxazole, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzoxazole, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 105680250
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.30 
Save 8.65 (14%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.30 
Save 8.65 (14%)
Add to cart
Benzoxazole, 97+%, Thermo Scientific Chemicals
Catalog number105680250
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.30 
Save 8.65 (14%)
-
Add to cart
Chemical Identifiers
CAS273-53-0
IUPAC Name1,3-benzoxazole
Molecular FormulaC7H5NO
InChI KeyBCMCBBGGLRIHSE-UHFFFAOYSA-N
SMILESO1C=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow
Appearance (Form)Low melting solid or liquid
Infrared spectrumConforms
Melting point27°C to 30°C
GC>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only