Thermo Scientific Chemicals

Benzoxazole, 97+%, Thermo Scientific Chemicals

Catalog number: 105681000
100 g, Each
Thermo Scientific Chemicals

Benzoxazole, 97+%, Thermo Scientific Chemicals

Catalog number: 105681000
100 g, Each
Quantity
Catalog number: 105681000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
273-53-0
IUPAC Name
1,3-benzoxazole
Molecular Formula
C7H5NO
InChI Key
BCMCBBGGLRIHSE-UHFFFAOYSA-N
SMILES
O1C=NC2=CC=CC=C12
Appearance (Color)
Colorless to white to yellow
Appearance (Form)
Low melting solid or liquid
Infrared spectrum
Conforms
Melting point
27°C to 30°C
GC
>=97.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text