Thermo Scientific Chemicals

Nalpha-Benzoyl-L-arginine ethyleester hydrochloride, ≥99%, Thermo Scientific Chemicals

Catalog number: 105700025
2.5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Nalpha-Benzoyl-L-arginine ethyleester hydrochloride, ≥99%, Thermo Scientific Chemicals

Catalog number: 105700025
2.5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 105700025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2645-08-1
IUPAC Name
hydrogen ethyl 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoate chloride
Molecular Formula
C15H23ClN4O3
InChI Key
HIXDELXKSSLIKB-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CCOC(=O)C(CCCN=C(N)N)NC(=O)C1=CC=CC=C1
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
TLC
>=99.0 %
Specific optical rotation
-15.0° to -19.0° (24°C, 589 nm) (c=2, water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text