Thermo Scientific Chemicals

4-Benzoylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 105720050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Benzoylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 105720050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 105720050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-95-0
IUPAC Name
4-benzoylbenzoic acid
Molecular Formula
C14H10O3
InChI Key
IFQUPKAISSPFTE-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Titration with NaOH
>=98.5 %
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
197°C to 200°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text