Thermo Scientific Chemicals

4-Benzoylbutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: 105740250
Each
Thermo Scientific Chemicals

4-Benzoylbutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: 105740250
Each
Catalog number: 105740250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1501-05-9
IUPAC Name
5-oxo-5-phenylpentanoic acid
Molecular Formula
C11H12O3
InChI Key
SHKWSBAVRQZYLE-UHFFFAOYSA-N
SMILES
OC(=O)CCCC(=O)C1=CC=CC=C1
Titration with NaOH
>=96.0 %
Appearance (Form)
Crystalline powder
Melting point
124°C to 128°C
Appearance (Color)
Off-white to light beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text