Thermo Scientific Chemicals

3-Benzoylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 105790250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Benzoylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 105790250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 105790250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2051-95-8
IUPAC Name
4-oxo-4-phenylbutanoic acid
Molecular Formula
C10H10O3
InChI Key
KMQLIDDEQAJAGJ-UHFFFAOYSA-N
SMILES
OC(=O)CCC(=O)C1=CC=CC=C1
Infrared spectrum
Conforms
Melting point
114°C to 119°C
Appearance (Color)
White to cream
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text