Gurr Buffer Tablets
Gurr Buffer Tablets
Gurr Buffer Tablets
Gibco™

Gurr Buffer Tablets

Gibco Gurr Buffer Tablets offer a convenient way to prepare a pH 6.8 phosphate buffer for dilution of Giemsa stainRead more
Have Questions?
Catalog NumberQuantity
1058201350 TabLets
Catalog number 10582013
Price (TWD)
3,990.00
Online offer
Ends: 31-Mar-2026
5,320.00
Save 1,330.00 (25%)
Each
Quantity:
50 TabLets
Price (TWD)
3,990.00
Online offer
Ends: 31-Mar-2026
5,320.00
Save 1,330.00 (25%)
Each
Gibco Gurr Buffer Tablets offer a convenient way to prepare a pH 6.8 phosphate buffer for dilution of Giemsa stain used in G-banding of chromosomes for cytogenetic analysis. Simply add one buffer tablet to 100 mL of distilled water and stir until dissolved.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
For Use With (Application)Clinical Research, Translational Research
Quantity50 TabLets
Shelf LifeSee product CoA
Shipping ConditionRoom Temperature
FormPowder
Product TypeGurr Buffer
pH6.8
Unit SizeEach
Contents & Storage
Storage conditions: 15–30°C
Shipping conditions: Ambient

Shelf life: 24 months from date of manufacture

Frequently asked questions (FAQs)

How long can I store Gurr Buffer Tablets (Cat. No. 10582013) that have been reconstituted?

After reconstituting Gurr Buffer Tablets, the subsequent solution can be stored at 4 degrees C for six months.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.