Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Thermo Scientific Chemicals

Benzyl benzoate, 99+%

CAS: 120-51-4 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Plastic drum
Catalog number 105860051
Price (USD)
204.00
Each
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
204.00
Each
Add to cart
Chemical Identifiers
CAS120-51-4
IUPAC Namebenzyl benzoate
Molecular FormulaC14H12O2
InChI KeySESFRYSPDFLNCH-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5670 to 1.5700 (20°C, 589 nm)
Acid value=<0.3 mg KOH/g
Appearance (Color)Clear colorless
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link