Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Thermo Scientific Chemicals

Benzyl benzoate, 99+%

CAS: 120-51-4 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 105862500
Price (USD)
43.65
Special offer
Online exclusive
Ends: 26-Sep-2025
53.70
Save 10.05 (19%)
Each
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 26-Sep-2025
53.70
Save 10.05 (19%)
Each
Add to cart
Chemical Identifiers
CAS120-51-4
IUPAC Namebenzyl benzoate
Molecular FormulaC14H12O2
InChI KeySESFRYSPDFLNCH-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5670 to 1.5700 (20°C, 589 nm)
Acid value=<0.3 mg KOH/g
Appearance (Color)Clear colorless
GC>=99.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection