Thermo Scientific Chemicals

Benzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 105871000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Benzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 105871000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 105871000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-39-0
IUPAC Name
(bromomethyl)benzene
Molecular Formula
C7H7Br
InChI Key
AGEZXYOZHKGVCM-UHFFFAOYSA-N
SMILES
BrCC1=CC=CC=C1
Refractive index
1.5745 to 1.5765 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text