Thermo Scientific Chemicals

Benzyl carbamate, 98%, Thermo Scientific Chemicals

Catalog number: 105890250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

Benzyl carbamate, 98%, Thermo Scientific Chemicals

Catalog number: 105890250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 105890250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
621-84-1
IUPAC Name
benzyl carbamate
Molecular Formula
C8H9NO2
InChI Key
PUJDIJCNWFYVJX-UHFFFAOYSA-N
SMILES
NC(=O)OCC1=CC=CC=C1
Appearance (Color)
Off-white to light beige
Melting point
86°C to 89°C
Appearance (Form)
Powder or flakes
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text