Benzyl carbamate, 98%, Thermo Scientific Chemicals
Benzyl carbamate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl carbamate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass Bottle
Catalog number 105891000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS621-84-1
IUPAC Namebenzyl carbamate
Molecular FormulaC8H9NO2
InChI KeyPUJDIJCNWFYVJX-UHFFFAOYSA-N
SMILESNC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
Melting point86°C to 89°C
Appearance (Form)Powder or flakes
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only