N-Benzylethanolamine, 96%, Thermo Scientific Chemicals
N-Benzylethanolamine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylethanolamine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 105945000
Price (USD)/ Each
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS104-63-2
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5420 to 1.5440 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only