Thermo Scientific Chemicals

Benzylhydrazine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 105960250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Benzylhydrazine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 105960250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 105960250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20570-96-1
IUPAC Name
dihydrogen benzylhydrazine dichloride
Molecular Formula
C7H12Cl2N2
InChI Key
MSJHOJKVMMEMNX-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC=C1
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 %
Appearance (Color)
Light yellow to beige
Appearance (Form)
Powder
Melting point
138°C to 143°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text