Invitrogen™

Exosome-Streptavidin Isolation/Detection Reagent

Catalog number: 10608D
Have Questions?
Invitrogen™

Exosome-Streptavidin Isolation/Detection Reagent

Catalog number: 10608D
Have Questions?
Catalog Number
10608D
Unit Size
3 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
10608D3 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Exosome-Streptavidin Isolation/Detection Reagent, when combined with your choice of biotinylated primary antibody, enables the purification of exosomes (also called extracellular vesicles and multi-vesicular bodies) from pre-enriched samples. These exosomes can then be detected using techniques such as flow cytometry, electron microscopy, or Western blotting. The exosomes must be pre-enriched before isolation. This can be done by ultracentrifugation, or by using the fast and efficient Total Exosome Isolation (from cell culture media) reagent.

• Maximum flexibility with your choice of biotinylated antibody
• 'See' your sample during handling
• Easily scalable protocol
• Detect exosomes via flow cytometry, in less than 1 hour

Isolation and detection of exosomes has been a tedious, non-specific, and difficult process. Exosome - Streptavidin for Isolation/Detection utilizes the well-known Dynabeads™ magnetic separation technology. When combined with the biotinylated antibody of your choice, this technology allows you to easily purify pre-enriched exosomes from cell culture media and then move on to detect the purified exosomes via techniques such as flow cytometry, electron microscopy, or Western blotting.

Detect by Flow Cytometry
Because free exosomes alone are too small to be detected by flow cytometry, one of the major advantages of employing magnetic separation technology is that the purified, bead-bound exosomes can be easily visualized by flow cytometry. The monodispersed and relatively large Dynabeads™ (4.5 μm in diameter) allow for a clear and defined FFC/SSC, typically in less than 1 hour.

'See' Your Sample
Not only are the superparamagnetic Dynabeads™ known for their sensitivity, reproducibility, and stability, but the magnetic handling also allows you to 'see' your sample due to the light brown color of the beads. When the sample tube is placed on the magnet, the bead-bound exosomes are pulled to the side of the tube, allowing for easy separation and purification. In addition, the sample and bead volume can be easily scaled up or down according to your sample size or downstream applications.

Good Mixing is Critical
For successful exosome isolation it is important to use a mixer that tilts and rotates to ensure that the beads do not settle in the tube. Avoid end-over end rotation for small sample volumes (e.g., 100 μl). Please see user manual below for more guidelines.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Cells in culture
Product Type
Isolation/Detection Reagent
Product Line
Dynabeads™
Sample Type
Cell Culture Supernatants
Target Species
Human
Diameter (Metric)
4.5 μm

Contents & Storage

Store at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text