5-Benzyloxytryptamine hydrochloride, 98%, Thermo Scientific Chemicals
5-Benzyloxytryptamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Benzyloxytryptamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 106110010
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
146.00 
Save 21.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
146.00 
Save 21.35 (15%)
Add to cart
5-Benzyloxytryptamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog number106110010
Price (USD)/ Each
124.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
146.00 
Save 21.35 (15%)
-
Add to cart
Chemical Identifiers
CAS52055-23-9
IUPAC Name2-[5-(benzyloxy)-1H-indol-3-yl]ethan-1-amine hydrochloridyl
Molecular FormulaC17H18ClN2O
InChI KeyNIDXFJIFFPKQBY-UHFFFAOYSA-N
SMILES[Cl].NCCC1=CNC2=CC=C(OCC3=CC=CC=C3)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow to beige
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only