Invitrogen™

Exosome-Human EpCAM Isolation Reagent (from cell culture)

Catalog number: 10618D
Have Questions?
Invitrogen™

Exosome-Human EpCAM Isolation Reagent (from cell culture)

Catalog number: 10618D
Have Questions?
Catalog Number
10618D
also known as 106-18D
Unit Size
2 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
10618D
also known as 106-18D
2 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Exosome-Human EpCAM Isolation Reagent (from cell culture) enables isolation of EpCAM-positive extracellular vesicles (ECVs) such as exosomes from enriched cell culture media. Following isolation, the specific sub-population of exosomes/ECVs can be further studied using methods like Western blotting, qPCR, or next generation sequencing.

• Minimal hands-on time for specific exosome isolation

• Obtain a highly pure exosome subset

• Protocol is easily scalable

• Compatible with most downstream analysis

Recent studies suggest that the exosome/ECV population consists of several subsets, possibly with different functions. Using the immuno-affinity based Dynabeads™ magnetic separation technology, these subsets can be selectively isolated and studied. The cell culture media must first be enriched for exosomes. This can be done quickly and efficiently using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation. Following enrichment, specific exosomes are isolated by binding to the surface of the magnetic beads.

Dynabeads™ magnetic beads are known for their sensitivity, reproducibility, and stability. Dynabeads™ magnetic separation technology also allows you to 'see' your sample during handling due to the color of the magnetic beads. When the sample tube is placed in the magnet, the bead-bound exosomes are pulled to the side of the tube, allowing for easy separation and buffer changes. In addition, the sample and bead volume can be easily scaled according to your sample size or downstream applications.

Note: It is important to provide sufficient mixing to ensure that the magnetic beads do not settle in the sample tube. We recommend using a mixer which provides both tilting and rotation for optimal mixing.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Isolation Reagent
Product Line
Dynabeads™
Sample Type
Cell Cultures

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text