Exosome-Human CD9 Flow Detection Reagent (from cell culture)
Exosome-Human CD9 Flow Detection Reagent (from cell culture)
Invitrogen™

Exosome-Human CD9 Flow Detection Reagent (from cell culture)

Exosome-Human CD9 Flow Detection Reagent (from cell culture) enables detection of CD9-positive extracellular vesicles (ECVs) such as exosomes in enrichedRead more
Have Questions?
Catalog NumberQuantity
10620D2 mL
Catalog number 10620D
Price (USD)
287.65
Online Exclusive
302.00
Save 14.35 (5%)
Each
-
Add to cart
Quantity:
2 mL
Price (USD)
287.65
Online Exclusive
302.00
Save 14.35 (5%)
Each
Add to cart
Ask our AI about this Product
Exosome-Human CD9 Flow Detection Reagent (from cell culture) enables detection of CD9-positive extracellular vesicles (ECVs) such as exosomes in enriched cell culture media using flow cytometry or electron microscopy. Free exosomes alone are too small to be detected by most flow cytometry instruments. By employing Dynabeads™ magnetic separation technology, exosomes can be easily visualized by flow cytometry while bound to the bead surface. The 2.7 μm Dynabeads™ magnetic beads allow for a clear and defined flow analysis.

• Minimal hands-on time for specific exosome capture and analysis

• Clear and defined flow analysis in less than 1 hour hands-on time

• Study expression of specific exosome subset markers

• Protocol is easily scalable

Recent studies suggest that the exosome/ECV population consists of several subsets, possibly with different functions. Using the immuno-affinity based Dynabeads™ magnetic separation technology, these subsets can be selectively captured and studied using flow cytometry and/or electron microscopy. And with Dynabeads™ technology you avoid losing material due to the handling and centrifugation typically experienced with other isolation and analysis methods.

The cell culture media must first be enriched for exosomes. This can be done quickly and efficiently using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation.

Note: It is important to provide sufficient mixing to ensure that the magnetic beads do not settle in the sample tube. We recommend using a mixer which provides both tilting and rotation for optimal mixing.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineDynabeads™
Quantity2 mL
Sample TypeCell Cultures
Diameter (Metric)2.7 μm
Product TypeFlow Detection Reagent
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3227200Certificate of AnalysisMay 22, 202510620D
3150179Certificate of AnalysisApr 30, 202510620D
2759045Certificate of AnalysisJan 29, 202510620D
2812140Certificate of AnalysisJan 29, 202510620D
2812142Certificate of AnalysisJan 28, 202510620D
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link