2-Biphenylcarboxylic acid, 98%, Thermo Scientific Chemicals
2-Biphenylcarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Biphenylcarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 106270250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS947-84-2
IUPAC Name[1,1'-biphenyl]-2-carboxylic acid
Molecular FormulaC13H10O2
InChI KeyILYSAKHOYBPSPC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point110°C to 114°C
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only