Thermo Scientific Chemicals

4-Biphenylcarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 106285000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Biphenylcarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 106285000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 106285000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92-92-2
IUPAC Name
[1,1'-biphenyl]-4-carboxylic acid
Molecular Formula
C13H10O2
InChI Key
NNJMFJSKMRYHSR-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Appearance (Color)
White to beige
Infrared spectrum
Conforms
Titration with NaOH
97.5 to 102.5 %
Melting point
223°C to 228°C
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text