Thermo Scientific Chemicals

2,2'-Biquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 106300050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2'-Biquinoline, 98%, Thermo Scientific Chemicals

Catalog number: 106300050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 106300050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
119-91-5
IUPAC Name
2,2'-biquinoline
Molecular Formula
C18H12N2
InChI Key
WPTCSQBWLUUYDV-UHFFFAOYSA-N
SMILES
C1=CC2=CC=C(N=C2C=C1)C1=NC2=CC=CC=C2C=C1
Appearance (Form)
Powder or crystals or flakes and/or chunks
Infrared spectrum
Conforms
Titration with HClO4
97.5 to 102.5 %
Appearance (Color)
White to cream to yellow to light brown
Melting point
192°C to 198°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text