Thermo Scientific Chemicals

Bis(2-chloroethyl)amine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 106321000
Each
Thermo Scientific Chemicals

Bis(2-chloroethyl)amine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 106321000
Each
Catalog number: 106321000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
821-48-7
IUPAC Name
bis(2-chloroethyl)azanium chloride
Molecular Formula
C4H10Cl3N
InChI Key
YMDZDFSUDFLGMX-UHFFFAOYSA-N
SMILES
[Cl-].ClCC[NH2+]CCCl
Appearance (Color)
White to beige
Melting point
210°C to 215°C
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text