Search Thermo Fisher Scientific
Handle reagents on automated systems simply and economically with Thermo Scientific™ Automation Reservoirs, which provide standard SBS dimensions ensuring compatibility with plate stackers and handlers. Reservoirs are disposable with no need for washing, allowing more reservoirs to be used per run. Built-in grooves in reservoir bottom reduce reagent waste and allow consistent aspiration.
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reservoir |
Includes | Removable splash baffles |
Channels | 384 |
Unit Size | Case of 20 |
Description | Deepwell |
Material | Plastic |
Sterility | Non-sterile |
Type | Reservoir |
Includes | Removable splash baffles |
Channels | 96 |
Unit Size | Case of 20 |
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reservoir |
Includes | Removable splash baffles |
Channels | 96 |
Unit Size | Case of 20 |
Description | Deepwell |
Material | Polypropylene |
Sterility | Sterile |
Type | Reservoir |
Includes | Removable splash baffles |
Packaging | Individually Wrapped |
Channels | 96 |
Unit Size | Case of 20 |
Material | Polypropylene |
Sterility | Sterile |
Type | Reservoir |
Includes | Removable splash baffles |
Packaging | Individually Wrapped |
Channels | 384 |
Unit Size | Case of 20 |
Material | Polypropylene |
Sterility | Sterile |
Type | Reservoir |
Includes | Removable splash baffles |
Packaging | Individually Wrapped |
Channels | 96 |
Unit Size | Case of 20 |
Catalog Number | Specifications | Unit Size | Channels | Sterility | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
1064-05-7 | Case of 20 | 384 | Non-sterile | 213.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1064-05-6 | Case of 20 | 96 | Non-sterile | 284.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1064-05-8 | Case of 20 | 96 | Non-sterile | 213.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1064-15-6 | Case of 20 | 96 | Sterile | 315.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1064-15-7 | Case of 20 | 384 | Sterile | 241.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1064-15-8 | Case of 20 | 96 | Sterile | 241.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Removable Splash Baffles: