Thermo Scientific Chemicals

Biuret, 97%, extra pure

Catalog number: 106521000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Biuret, 97%, extra pure

Catalog number: 106521000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 106521000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-19-0
IUPAC Name
1-(carbamoylamino)formamide
Molecular Formula
C2H5N3O2
InChI Key
OHJMTUPIZMNBFR-UHFFFAOYSA-N
SMILES
NC(=O)NC(N)=O
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Melting point
184°C to 190°C
Appearance (Color)
White
Assay
>=96.0 % (ex N)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text