Thermo Scientific Chemicals

Bromoacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 106570100
10 kg, Plastic Pail, Each
Thermo Scientific Chemicals

Bromoacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 106570100
10 kg, Plastic Pail, Each
Quantity
Packaging
Catalog number: 106570100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-08-3
IUPAC Name
2-bromoacetic acid
Molecular Formula
C2H3BrO2
InChI Key
KDPAWGWELVVRCH-UHFFFAOYSA-N
SMILES
OC(=O)CBr
Appearance (Form)
and/or chunks
Titration with NaOH
98.0 to 102.0 %
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals or flakes or mass

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text