2-Bromoaniline, 98%, Thermo Scientific Chemicals
2-Bromoaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromoaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 106611000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS615-36-1
IUPAC Name2-bromoaniline
Molecular FormulaC6H6BrN
InChI KeyAOPBDRUWRLBSDB-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear yellow to red-brown
GC>=97.5 %
Appearance (Form)Liquid after melting
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only