4-Bromoanisole, 98%, Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromoanisole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 106631000
Price (USD)/ Each
78.65
Online exclusive
87.60 
Save 8.95 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
78.65
Online exclusive
87.60 
Save 8.95 (10%)
Add to cart
4-Bromoanisole, 98%, Thermo Scientific Chemicals
Catalog number106631000
Price (USD)/ Each
78.65
Online exclusive
87.60 
Save 8.95 (10%)
-
Add to cart
Chemical Identifiers
CAS104-92-7
IUPAC Name1-bromo-4-methoxybenzene
Molecular FormulaC7H7BrO
InChI KeyQJPJQTDYNZXKQF-UHFFFAOYSA-N
SMILESCOC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5630 to 1.5650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only