Thermo Scientific Chemicals

2-Bromobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 106650100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 106650100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 106650100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6630-33-7
Appearance (Form)
Liquid after melting
Appearance (Color)
Clear yellow
Refractive index
1.5950 to 1.5970 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text