Thermo Scientific Chemicals

3-Bromobenzaldehyde, 96%, Thermo Scientific Chemicals

Catalog number: 106660250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Bromobenzaldehyde, 96%, Thermo Scientific Chemicals

Catalog number: 106660250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 106660250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3132-99-8
IUPAC Name
3-bromobenzaldehyde
Molecular Formula
C7H5BrO
InChI Key
SUISZCALMBHJQX-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(C=O)=C1
Refractive index
1.5920 to 1.5960 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid after melting
GC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text