Appearance (Form)Crystalline powder or crystals or crystalline mass
Melting point37°C to 41°C
Appearance (Color)White to light yellow to beige
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.