4-Bromobenzophenone, 97%, Thermo Scientific Chemicals
4-Bromobenzophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromobenzophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 106721000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS90-90-4
IUPAC Name(4-bromophenyl)(phenyl)methanone
Molecular FormulaC13H9BrO
InChI KeyKEOLYBMGRQYQTN-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point78°C to 82°C
GC>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only