Search Thermo Fisher Scientific
Choose from this range of sample and skimmer cones to suit your analysis requirements when using the Thermo Scientific™ ELEMENT™ Series of HR-ICP-MS instruments.
Catalog Number | Type | Material |
---|---|---|
1067500 | Sample Cone | Platinum, Boron-free |
1044530 | Sample Cone | Nickel |
1047460 | Skimmer Cone | Platinum, Boron-free |
1047461 | Skimmer Cone | Platinum |
1067501 | Sample Cone | Platinum |
1067600 | Skimmer Cone | Nickel |
1142150 | Skimmer Cone | Platinum, X-Vers, Boron-free |
1142151 | Skimmer Cone | Platinum, X-Version |
1142160 | Skimmer Cone | Nickel, X-Version |
1184050 | Sample Cone | Aluminum |
1184060 | Skimmer Cone | Aluminum |
1260630 | Jet Sample Cone | Nickel |
BRE0010791 | Jet Sample Cone | Platinum, Boron-free |
BRE0010792 | Jet Sample Cone | Platinum w/ Boron |
A range of easily changeable Ni, Pt, and Al Thermo Scientific™ Sample and Skimmer Cones are available for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS and Thermo Scientific™ Neptune™ Series MC-ICP-MS systems to suit your analysis requirements, all designed for durability, performance and ease of use.
Select High-Quality Consumables for Consistent Performance
These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in solid Ni construction, Pt-tipped for even greater durability, and Al for specific analysis requirements.
Share catalog number, name or link