4-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals
4-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 106760100
Price (USD)/ Each
41.50
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
41.50
Add to cart
4-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals
Catalog number106760100
Price (USD)/ Each
41.50
-
Add to cart
Chemical Identifiers
IUPAC Name4-bromobenzoyl chloride
Molecular FormulaC7H4BrClO
InChI KeyDENKGPBHLYFNGK-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=C(Br)C=C1
Molecular Weight (g/mol)219.46
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Infrared spectrumConforms
Appearance (Form)Low melting solid
Titration RCOCl>=97.5 %
Melting point36°C to 41°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only