Thermo Scientific Chemicals

4-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 106761000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 106761000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 106761000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
586-75-4
IUPAC Name
4-bromobenzoyl chloride
Molecular Formula
C7H4BrClO
InChI Key
DENKGPBHLYFNGK-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(Br)C=C1
Appearance (Color)
White to light brown
Infrared spectrum
Conforms
Appearance (Form)
Low melting solid
Titration RCOCl
>=97.5 %
Melting point
36°C to 41°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text