1-Bromo-4-chlorobutane, 99%, Thermo Scientific Chemicals
1-Bromo-4-chlorobutane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-4-chlorobutane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 106840250
Price (USD)/ Each
38.65
Online exclusive
42.80 
Save 4.15 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
38.65
Online exclusive
42.80 
Save 4.15 (10%)
Add to cart
1-Bromo-4-chlorobutane, 99%, Thermo Scientific Chemicals
Catalog number106840250
Price (USD)/ Each
38.65
Online exclusive
42.80 
Save 4.15 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-bromo-4-chlorobutane
Molecular FormulaC4H8BrCl
InChI KeyNIDSRGCVYOEDFW-UHFFFAOYSA-N
SMILESClCCCCBr
Molecular Weight (g/mol)171.46
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4865 to 1.4885 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only