1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals
1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 106860025
Price (USD)/ Each
247.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
291.00 
Save 43.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
247.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
291.00 
Save 43.35 (15%)
Add to cart
1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals
Catalog number106860025
Price (USD)/ Each
247.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
291.00 
Save 43.35 (15%)
-
Add to cart
Chemical Identifiers
CAS109-70-6
IUPAC Name1-bromo-3-chloropropane
Molecular FormulaC3H6BrCl
InChI KeyMFESCIUQSIBMSM-UHFFFAOYSA-N
SMILESClCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4850 to 1.4880 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only