Hygromycin B (50 mg/mL)
Gibco™

Hygromycin B (50 mg/mL)

Hygromycin B is a water-soluble antibiotic purified from the bacterium Streptomyces hydroscopicus. Gibco™ Hygromycin B is used as a bacterialRead more
Have Questions?
Catalog NumberQuantity
1068701020 mL
Catalog number 10687010
Price (USD)
269.00
Each
Quantity:
20 mL
Price (USD)
269.00
Each
Hygromycin B is a water-soluble antibiotic purified from the bacterium Streptomyces hydroscopicus. Gibco™ Hygromycin B is used as a bacterial selection antibiotic in the concentration range of 200–500 μg/mL. This product is supplied as a 50 mg/mL solution.

Resistance to Hygromycin B is conferred by the E. coli hygromycin resistance gene (hyg or hph). Hygromycin B has a different mode of action than Geneticin™, Blasticidin S, or Zeocin™ selection reagents, making it an excellent choice for dual-selection experiments (see selection antibiotics).

Other Choices and More Information
We offer a wide range of antibiotics and antimycotics in both powder and liquid formats.

See the complete list, or find products for:
Contamination control
Eukaryotic and bacterial selection

See recommendations for working concentrations for selection antibiotics.

Learn more about the use of antibiotics and antimycotics in cell culture, and review guidelines for decontaminating cultures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration50 mg/mL
Culture TypeMammalian Cell Culture, Insect Cell Culture
For Use With (Application)Eukaryotic Selection⁄Stable Cell Line Generation
Quantity20 mL
Shipping ConditionRoom Temperature
FormLiquid
Product TypeAntibiotic
SterilitySterile
Unit SizeEach
Contents & Storage
Storage conditions: 2 to 8°C
Shipping conditions: Ambient

Frequently asked questions (FAQs)

Which of your antibiotics (Geneticin, Zeocin, Hygromycin B, Blasticidin, and Puromycin) can be used together for stable selection in mammalian cells?

All of our antibiotics (Geneticin, Zeocin, Hygromycin B, Blasticidin, and Puromycin) can be used together for making multiple stable cell lines. However, kill curves will need to be performed for each combination of antibiotics since sensitivity to a given antibiotic tends to increase when combined with other antibiotics.

What are the recommended concentrations of antibiotics to use for selection in prokaryotes and eukaryotes?

For best results, optimal concentrations for selection should be determined empirically in each unique experiment through dose response curves. However, to get a general idea of concentrations that have worked for individual cell types, please click on the following url: http://www.thermofisher.com/us/en/home/life-science/cell-culture/transfection/selection.html or type in “Selection Antibiotics” into our main search on www.thermofisher.com.

What is the range of specific activity of Hygromycin B?

The specific activity range of Hygromycin B is from 386-482 micrograms/mg.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What solvent is used to prepare your Hygromycin B solution? How should it be stored?

Hygromycin B is supplied as a 50 mg/mL solution in phosphate buffered saline. It is manufactured aseptically and sterile filtered, and is suitable for use in tissue culture applications. We recommend storing it at 2-8 degrees C.

What is the mode of action on the following antibiotics: Blasticidin, Geneticin (G418), Hygromycin, and Zeocin?

Blasticidin: Nucleoside Inhibits protein synthesis in prokaryotic and eukaryotic cells by interfering with peptidyl transfer reaction of protein synthesis, causing early termination of translation.

Geneticin (G418): Aminoglycoside Blocks protein synthesis in mammalian cells by interfering with ribosomal function.

Hygromycin: Aminocyclitol Inhibits protein synthesis by disrupting translocation and promoting mistranslation.

Zeocin: Intercalates with DNA and cleaves it.

Citations & References (38)

Citations & References
Abstract
WAF1, a potential mediator of p53 tumor suppression.
Authors:el-Deiry WS, Tokino T, Velculescu VE, Levy DB, Parsons R, Trent JM, Lin D, Mercer WE, Kinzler KW, Vogelstein B
Journal:Cell
PubMed ID:8242752
The ability of p53 to activate transcription from specific sequences suggests that genes induced by p53 may mediate its biological role as a tumor suppressor. Using a subtractive hybridization approach, we identified a gene, named WAF1, whose induction was associated with wild- type but not mutant p53 gene expression in ... More
CC chemokine receptor 5-mediated signaling and HIV-1 Co-receptor activity share common structural determinants. Critical residues in the third extracellular loop support HIV-1 fusion.
Authors:Alkhatib G, Ahuja SS, Light D, Mummidi S, Berger EA, Ahuja SK
Journal:J Biol Chem
PubMed ID:9242636
'There is a close correspondence between the ability of RANTES and macrophage inflammatory proteins 1alpha and 1beta to activate CC chemokine receptor 5 (CCR5) and the ability to inhibit CCR5-dependent membrane fusion mediated by the envelope glycoprotein of human immunodeficiency virus (HIV), type 1. This finding suggests that some of ... More
Dimerization of a selectin and its ligand stabilizes cell rolling and enhances tether strength in shear flow.
Authors: Ramachandran V; Yago T; Epperson T K; Kobzdej M M; Nollert M U; Cummings R D; Zhu C; McEver R P;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11526223
'Selectins mediate rolling of leukocytes by rapid formation and dissociation of selectin-ligand bonds, which are assumed to require high mechanical strength to prevent premature dissociation by the forces applied in shear flow. This assumption is based largely on the observation that increasing wall shear stress increases only modestly the dissociation ... More
Reconstitution of stretch-activated cation channels by expression of the alpha-subunit of the epithelial sodium channel cloned from osteoblasts [published erratum appears in Proc Natl Acad Sci U S A 1997 Apr 15;94(8):4233]
Authors:Kizer N, Guo XL, Hruska K
Journal:Proc Natl Acad Sci U S A
PubMed ID:9023374
'Osteoblasts respond to repetitive strain by activating stretch- activated, nonselective cation channels (SA-CAT) and increasing matrix protein production. SA-CAT channels are thought to be responsible for mechano-transduction in osteoblasts, although the molecular identity of the SA-CAT channel has previously been unknown. We have demonstrated that both the UMR-106 osteoblast-like cell ... More
Genetic analysis of the herpes simplex virus type 1 UL20 protein domains involved in cytoplasmic virion envelopment and virus-induced cell fusion.
Authors:Melancon JM, Foster TP, Kousoulas KG,
Journal:J Virol
PubMed ID:15220406
'The herpes simplex virus type 1 UL20 protein (UL20p) is an important determinant for cytoplasmic virion morphogenesis and virus-induced cell fusion. To delineate the functional domains of the UL20 protein, we generated a panel of single and multiple (cluster) alanine substitutions as well as UL20p carboxyl-terminal truncations. The UL20 mutant ... More