Thermo Scientific Chemicals

4-Bromo-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 106875000
500 g, Each
Thermo Scientific Chemicals

4-Bromo-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 106875000
500 g, Each
Quantity
Catalog number: 106875000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7463-51-6
IUPAC Name
4-bromo-3,5-dimethylphenol
Molecular Formula
C8H9BrO
InChI Key
WMUWDPLTTLJNPE-UHFFFAOYSA-N
SMILES
CC1=CC(O)=CC(C)=C1Br
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Color)
Colorless to light pink
Melting point
113°C to 116°C
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text