Thermo Scientific Chemicals

1-Bromododecane, 98%, Thermo Scientific Chemicals

Catalog number: 106910010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromododecane, 98%, Thermo Scientific Chemicals

Catalog number: 106910010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 106910010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
143-15-7
IUPAC Name
1-bromododecane
Molecular Formula
C12H25Br
InChI Key
PBLNBZIONSLZBU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCBr
Color scale
=<125 APHA
Infrared spectrum
Conforms
Appearance (Form)
Clear liquid
GC
>=97.5 %
Refractive index
1.4570 to 1.4590 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text