Thermo Scientific Chemicals

2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals

Catalog number: 106931000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals

Catalog number: 106931000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 106931000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2576-47-8
IUPAC Name
hydrogen 2-bromoethan-1-amine bromide
Molecular Formula
C2H7Br2N
InChI Key
WJAXXWSZNSFVNG-UHFFFAOYSA-N
SMILES
[H+].[Br-].NCCBr
Appearance (Form)
Crystalline powder or crystals and/or chunks
Titration Argentometric
>=98.5 %
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Melting point
172°C to 176°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text