2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 106932500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namehydrogen 2-bromoethan-1-amine bromide
Molecular FormulaC2H7Br2N
InChI KeyWJAXXWSZNSFVNG-UHFFFAOYSA-N
SMILES[H+].[Br-].NCCBr
Molecular Weight (g/mol)204.89
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Argentometric>=98.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point172°C to 176°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only