(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals
(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 106941000
Price (USD)/ Each
46.30
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.30
Add to cart
(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals
Catalog number106941000
Price (USD)/ Each
46.30
-
Add to cart
Chemical Identifiers
CAS103-63-9
IUPAC Name(2-bromoethyl)benzene
Molecular FormulaC8H9Br
InChI KeyWMPPDTMATNBGJN-UHFFFAOYSA-N
SMILESBrCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5553 to 1.5573 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Water=<0.05 %
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only