Thermo Scientific Chemicals

2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 106990500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 106990500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 106990500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1072-85-1
IUPAC Name
1-bromo-2-fluorobenzene
Molecular Formula
C6H4BrF
InChI Key
IPWBFGUBXWMIPR-UHFFFAOYSA-N
SMILES
FC1=CC=CC=C1Br
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.5327 to 1.5347 (20°C, 589 nm)
GC
>=98.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text