Thermo Scientific Chemicals

1-Bromonaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: 107150025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromonaphthalene, 96%, Thermo Scientific Chemicals

Catalog number: 107150025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 107150025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-11-9
IUPAC Name
1-bromonaphthalene
Molecular Formula
C10H7Br
InChI Key
DLKQHBOKULLWDQ-UHFFFAOYSA-N
SMILES
BrC1=C2C=CC=CC2=CC=C1
GC
>=95 %
Appearance (Color)
Clear yellow to yellow-brown
Infrared spectrum
Conforms
GC
=<2.5 % 2-Bromonaphthalene
Refractive index
1.6560 to 1.6590 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text